missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p21 (Thr145) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538144
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Protein p21 is a tumor suppressor protein which is an inhibitor of Cyclin-dependent kinases (CDK) and is transcriptionally activated by p53. P21 is important in the response of cells to genotoxic stress and a major transcriptional target of p53 protein. The occurrence of p21 in the nucleus executes binding and inhibition activity of cyclin dependent kinases Cdk1 and Cdk2, and blocks the transition from G1 phase into S phase or from G2 phase into mitosis after DNA damage, which enables the repair of damaged DNA. In the cytoplasm, p21 protein has an anti-apoptotic effect. P21 is able to bind to and inhibit caspase 3, as well as the apoptotic kinases ASK1 and JNK. P21 exhibits a dual function in carcinogenesis, and acts as a tumor suppressor, prevents apoptosis, and acts as an oncogene.Spezifikation
Phospho-p21 (Thr145) | |
Polyclonal | |
Unconjugated | |
CDKN1A | |
Cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, Melanoma differentiation-associated protein 6, MDA-6, CDKN1A, CAP20, CDKN1, CIP1, MDA6, PIC1, SDI1, WAF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1026 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P38936 | |
CDKN1A | |
A synthetic phosphopeptide derived from human p21Cip1 around the phosphorylation site of Thr145 (R-Q-TP-S-M) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |