missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p21 (Thr57) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA512647
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Phospho-p21 (Thr57) Polyclonal antibody specifically detects Phospho-p21 (Thr57) in Human samples. It is validated for Dot Blot, Immunohistochemistry.Spezifikation
Phospho-p21 (Thr57) | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P38936 | |
CDKN1A | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T57 of human P21CIP1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Dot Blot, Immunohistochemistry | |
Unconjugated | |
CDKN1A | |
Activating Fragment 1; CAP20; cdk inhibitor CIP1; cdk inhibitor CIP1 (p21); CDKI; CDK-interacting protein 1; CDK-interaction protein 1; Cdkn1; CDKN1A; CDNK1A; CIP1; Cip1); cyclin dependent kinase inhibitor 1A; cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A; Cyclin-dependent kinase inhibitor 1A (p21; cyclin-dependent kinase inhibitor 1A (P21); cyclin-dependent kinase inhibitor 1A (p21, Cip1); cyclin-dependent kinase inhibitor 1A variant 1; cyclin-dependent kinase inhibitor 1A variant 2; DNA synthesis inhibitor; MDA6; MDA-6; melanoma differentiation associated protein 6; melanoma differentiation-associated protein; Melanoma differentiation-associated protein 6; OTTHUMP00000016298; p21; p21<WAF1/CIP1>; p21Cip1; p21Cip1/Waf1; p21WAF; PIC1; putative cyclin-dependent kinase inhibitor 1A (p21/WAF1/CIP1); SDI1; SLC12A9; UV96; WAF1; Wild type p53 activated fragment 1 (WAF1); wild-type p53-activated fragment 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1026 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |