missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p38 MAPK alpha (Thr180, Tyr182) Monoclonal Antibody (F.52.8), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA515177
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the phosphorylated forms of either p42/44 MAPK or SAPK/JNK.
Phospho-p38 MAPK alpha (Thr180, Tyr182) Monoclonal antibody specifically detects Phospho-p38 MAPK alpha (Thr180, Tyr182) in Drosophila, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Phospho-p38 MAPK alpha (Thr180, Tyr182) | |
Monoclonal | |
71 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
O61443, P47811, P70618, Q16539 | |
MAPK14, p38b | |
Synthetic phosphopeptide corresponding to residues surrounding pThr180/Tyr182 of human p38 MAPK | |
200 μL | |
Primary | |
Drosophila, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
F.52.8 | |
Unconjugated | |
MAPK14 | |
186F5S; anon-sts23; BG:DS00797.3; CG7393; CG7393-PA; CRK1; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; Csbp2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; D-p38; D-p38 MAPK; Dp38b; D-p38b; EC 2.7.11.24; ERK1; ERT2; ESTS:186F5S; etID18993.8; Exip; fk28c03; hm:zeh1243; Hog; I79_016605; LD24658p; MAP 14; MAP kinase 1; MAP kinase 11; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38a; MAP kinase p38alpha; MAP kinase p38b; MAPK; MAPK 11; MAPK 14; MAPK 14A; MAPK1 antibody; Mapk11; Mapk14; mapk14a; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen | |
Rabbit | |
Affinity chromatography | |
RUO | |
1432, 26416, 34780, 81649 | |
-20°C | |
Liquid |