Learn More
Phospho-p38 MAPK (Thr180, Tyr182) Monoclonal Antibody (S.417.1), Invitrogen™
Rabbit Monoclonal Antibody
Marke:  Invitrogen MA515182
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11529501
  • 466.00 € / 200 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay. It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the phosphorylated forms of either p42/44 MAPK or SAPK/JNK.
Phospho-p38 MAPK (Thr180, Tyr182) Monoclonal antibody specifically detects Phospho-p38 MAPK (Thr180, Tyr182) in Drosophila, Human, Mouse, Non-human Primate, Porcine, Rat, Yeast samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
Spezifikation
Spezifikation
Phospho-p38 MAPK (Thr180, Tyr182)
Monoclonal
3 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
O61443, P47811, P70618, Q16539
MAPK14, p38b
Synthetic phosphopeptide corresponding to residues surrounding pThr180/Tyr182 of human p38 MAPK
200 μL
Primary
Drosophila, Human, Mouse, Non-human Primate, Porcine, Rat, Yeast
Antibody
IgG
Flow Cytometry, Immunocytochemistry, Western Blot
S.417.1
Unconjugated
MAPK14
186F5S; anon-sts23; BG:DS00797.3; CG7393; CG7393-PA; CRK1; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; Csbp2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; D-p38; D-p38 MAPK; Dp38b; D-p38b; EC 2.7.11.24; ERK1; ERT2; ESTS:186F5S; etID18993.8; Exip; fk28c03; hm:zeh1243; Hog; I79_016605; LD24658p; MAP 14; MAP kinase 1; MAP kinase 11; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38a; MAP kinase p38alpha; MAP kinase p38b; MAPK; MAPK 11; MAPK 14; MAPK 14A; MAPK1 antibody; Mapk11; Mapk14; mapk14a; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen
Rabbit
Affinity chromatography
RUO
100156630, 1432, 26416, 34780, 81649
-20°C