Learn More
Phospho-ERK1/ERK2 (Thr202, Tyr204) Monoclonal Antibody (S.812.9), Invitrogen™
Rabbit Monoclonal Antibody
Marke:  Invitrogen MA515173
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11372362
  • 473.00 € / 200 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the corresponding phosphorylated residues of either JNK/SAPK or p38 MAP kinase.
Phospho-ERK1/ERK2 (Thr202, Tyr204) Monoclonal antibody specifically detects Phospho-ERK1/ERK2 (Thr202, Tyr204) in Drosophila, Hamster, Human, Mink, Mouse, Non-human Primate, Porcine, Rat, Yeast, Zebrafish samples. It is validated for ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation
Spezifikation
Phospho-ERK1/ERK2 (Thr202, Tyr204)
Monoclonal
52.0 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P21708, P27361, P28482, P40417, P63085, P63086, Q63844
MAPK1, MAPK3, rl
Synthetic phosphopeptide corresponding to residues surrounding pThr202/Tyr204 of human p44 MAP kinase
200 μL
Primary
Drosophila, Hamster, Human, Mink, Mouse, Non-human Primate, Porcine, Rat, Yeast, Zebrafish
Antibody
IgG
ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S.812.9
Unconjugated
MAPK3
12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupI
Rabbit
Affinity chromatography
RUO
100153927, 100753149, 100759514, 116590, 26413, 26417, 3354888, 360144, 399480, 445013, 50689, 5594, 5595
-20°C