missing translation for 'onlineSavingsMsg'
Learn More

Phospho-NFkB p50 (Ser337) Recombinant Rabbit Monoclonal Antibody (2H7L8), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen 701430

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15480624

  • 468.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

701430 has been successfully used in Western blot, flow cytometry, and immunofluorescence applications. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Phospho-NFkB p50 (Ser337) Recombinant Monoclonal antibody specifically detects Phospho-NFkB p50 (Ser337) in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Phospho-NFkB p50 (Ser337)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
Rabbit
Protein A
RUO
Human
Liquid
Immunocytochemistry, Western Blot
2H7L8
Unconjugated
CVID12; DKFZp686C01211; DNA-binding factor KBF1; EBP-1; KBF1; MGC54151; NF kappaB1; NF KB; NFkappaB; NF-kappaB; NF-kappa-B; NF-kappaB p50; NF-kappaB1; NF-kappa-B1 p84/NF-kappa-B1 p98; NF-kappabeta; NF-kB; NFKB1; NF-kB1; NFKB-p105; NFKB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit 1; nuclear factor kappa-B 1; nuclear factor kappa-B DNA binding subunit; nuclear factor kappaB p50; nuclear factor NF-kappa-B p105 subunit; nuclear factor NF-kappa-B p105 subunit-like protein; nuclear factor NF-kappa-B p50 subunit; nuclear factor of kappa light chain gene enhancer in B-cells 1, p105; nuclear factor of kappa light polypeptide enhancer in B-cells 1; nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105; nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells 1 (p105); nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105; nuclear factor-kappaB
Phosphorylation of the p50 subunit at the Ser-337 residue
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote