missing translation for 'onlineSavingsMsg'
Learn More

Phospho-p53 (Ser15) Monoclonal Antibody (C.381.0), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA515229

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11599541

  • 469.00 € / 100 Mikroliter
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with p53 phosphorylated at other sites.

Phospho-p53 (Ser15) Monoclonal antibody specifically detects Phospho-p53 (Ser15) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Phospho-p53 (Ser15)
Monoclonal
765 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P04637
Tp53
Synthetic phosphopeptide corresponding to residues surrounding pSer15 of human p53
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Western Blot
C.381.0
Unconjugated
Tp53
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3
Mouse
Affinity chromatography
RUO
7157
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote