missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p53 (Ser392) Monoclonal Antibody (FP3.2 [FPS392]), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA119424
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For IHC staining, high temperature antigen retrieval (microwave, pressure cooker) is recommended, using 10 mM citrate buffer, pH 6.0, or 1 mM EDTA-NaOH buffer, pH 8.0.
Phospho-p53 (Ser392) Monoclonal antibody specifically detects Phospho-p53 (Ser392) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Phospho-p53 (Ser392) | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P04637 | |
Tp53 | |
KLH-conjugated phosphopeptide RHKKLMFKTEGPDS[P]D, corresponding to amino acids 378-393 of human p53. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
FP3.2 [FPS392] | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
Protein A | |
RUO | |
7157 | |
4° C, do not freeze | |
Liquid |