missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-VPS34 (Ser676) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA512799
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse, porcine, rat and Xenopus based on sequence homology.
Phospho-VPS34 (Ser676) Polyclonal antibody specifically detects Phospho-VPS34 (Ser676) in Human samples. It is validated for Dot Blot.Spezifikation
Phospho-VPS34 (Ser676) | |
Polyclonal | |
Unconjugated | |
Pik3c3 | |
5330434F23Rik; catalytic phosphatidylinositol 3-kinase 3; catalytic subunit type 3 Antibody; class 3 phosphoinositide-3-kinase; Class III PI 3-kinase; hVps34; hVps34 Antibody; OTTMUSP00000023341; OTTMUSP00000023350; phosphatidylinositol 3-kinase; Phosphatidylinositol 3-kinase catalytic subunit type 3; phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI3K type 3; PI-3K Vps34p; pi3kc3; PI3-kinase type 3; Pik3c3; PIK3C3 Antibody; PtdIns-3-kinase type 3; VPS34 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot | |
0.32 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NEB9 | |
Pik3c3 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S676 of human PI3KC3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |