Learn More
Phospho-PKC mu (Ser744, Ser748) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA517864
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11537110
  • 473.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
It is not recommended to aliquot this antibody.
Phospho-PKC mu (Ser744, Ser748) Polyclonal antibody specifically detects Phospho-PKC mu (Ser744, Ser748) in Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.
Spezifikation
Spezifikation
Phospho-PKC mu (Ser744, Ser748)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
O94806, Q15139, Q5XIS9, Q62101, Q8BZ03, Q8K1Y2, Q9BZL6, Q9WTQ1
PRKD1, Prkd2, Prkd3
Synthetic phosphopeptide corresponding to residues surrounding pSer744/748 of mouse PKD
100 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG
Western Blot
Unconjugated
Prkd2
4930557O20Rik; 5730497N19Rik; AI325941; DKFZP586E0820; EPK2; HSPC187; nPKC-D1; nPKC-D2; nPKC-mu; nPKC-nu; PKC mu; PKC nu; PKC u; PKC v; Pkcm; PKCmu; PKC-MU; Pkcnu; PKC-NU; PKCu; PKC-u; PKCv; PKC-v; Pkd; PKD1; PKD2; PKD3; Prkcm; PRKCN; Prkd1; PRKD2; PRKD3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D2; protein kinase D3; protein kinase EPK2; RGD1308054; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D2; Serine/threonine-protein kinase D3
Rabbit
Antigen affinity chromatography
RUO
101540, 18760, 23683, 25865, 292658, 313834, 5587, 75292, 85421
-20°C