missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-SSB (Ser366) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538816
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU(OH) elements of newly RNA polymerase III (see MIM 606007)-transcribed RNA, processing 5-prime and 3-prime ends of pre-tRNA precursors, acting as an RNA chaperone, and binding viral RNAs associated with hepatitis C virus. La protein was originally defined by its reactivity with autoantibodies from patients with Sjogren syndrome (MIM 270150) and systemic lupus erythematosus (SLE; MIM 152700) (Teplova et al., 2006 [PubMed 16387655]).Spezifikation
Phospho-SSB (Ser366) | |
Polyclonal | |
Unconjugated | |
SSB | |
LARP3, La, La ribonucleoprotein domain family, member 3, SS-B, SS-B/La protein, autoantigen La, la autoantigen, lupus La antigen, lupus La protein, sjoegren syndrome type B antigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6741 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05455 | |
SSB | |
A synthetic phosphopeptide derived from human SSB around the phosphorylation site of Ser366 (F-A-SP-D-D) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |