missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5 beta (Ser731) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538093
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Stat 5a protein is a transcription factor activated by hormone and cytokines. Two highly related, but distinct Stat 5 genes (Stat 5a and Stat 5b) were identified in mouse. The amino acid sequences of Stat 5a and Stat 5b show 96% sequence similarity, and both proteins are co expressed in most tissues of both virgin and lactating mice. However, differential accumulation of Stat 5a and Stat 5b mRNA has been reported for both muscle and mammary tissue. Stat 5a is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.Spezifikation
Phospho-STAT5 beta (Ser731) | |
Polyclonal | |
Unconjugated | |
STAT5B | |
Signal transducer and activator of transcription 5B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20851, 25126, 6777 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P51692, P42232, P52632 | |
STAT5B | |
A synthetic phosphopeptide derived from human STAT5B around the phosphorylation site of Ser731 (A-P-SP-P-A) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |