missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tau (Ser199) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 44734G
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Phospho-Tau (Ser199) Polyclonal antibody specifically detects Phospho-Tau (Ser199) in Human, Mouse, Rat samples. It is validated for Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-Tau (Ser199) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P10636, P10637, P19332 | |
MAPT | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Tau that contains serine 199. The sequence is conserved in mouse and rat. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
MAPT | |
AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17762, 29477, 4137 | |
-20°C | |
Liquid |