missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TSC2 (Ser1798) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA512908
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Phospho-TSC2 (Ser1798) Polyclonal antibody specifically detects Phospho-TSC2 (Ser1798) in Human samples. It is validated for Dot Blot, Immunocytochemistry.Spezifikation
Phospho-TSC2 (Ser1798) | |
Polyclonal | |
Unconjugated | |
TSC2 | |
LAM; Nafld; OTTHUMP00000198394; PPP1R160; protein phosphatase 1, regulatory subunit 160; Rc; renal carcinoma; Tcs2; Tsc2; TSC4; Tuberin; tuberous sclerosis 2; tuberous sclerosis 2 homolog protein; tuberous sclerosis 2 protein; Tuberous sclerosis 2 protein homolog | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
7249 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot, Immunocytochemistry | |
0.47 mg/mL | |
PBS with 0.09% sodium azide | |
P49815 | |
TSC2 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S1798 of human TSC2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |