missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TYK2 (Tyr1054, Tyr1055) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517898
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Tyk2 protein phosphorylated at other sites.
Phospho-TYK2 (Tyr1054, Tyr1055) Polyclonal antibody specifically detects Phospho-TYK2 (Tyr1054, Tyr1055) in Human samples. It is validated for Western Blot.Spezifikation
Phospho-TYK2 (Tyr1054, Tyr1055) | |
Polyclonal | |
Unconjugated | |
TYK2 | |
4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; IMD35; JTK1; mKIAA4116; Non-receptor tyrosine-protein kinase TYK2; Tyk2; tyrosine kinase 2; tyrosine kinase TYK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7297 | |
-20°C | |
Liquid |
Western Blot | |
121 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P29597 | |
TYK2 | |
Synthetic phosphopeptide corresponding to residues surrounding pTyr1054/1055 of human Tyk2 protein | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |