Learn More
Phospholipase Cg (pY783) Mouse, Unlabeled, Clone: 27, BD
Mouse Monoclonal Antibody
308.08 € - 724.00 €
Spezifikation
Antigen | Phospholipase Cg (pY783) |
---|---|
Klon | 27 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15886189
|
BD Biosciences
612464 |
50 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
15896189
|
BD Biosciences
612465 |
150 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
The Phospholipase C (PLC) isozymes hydrolyze phosphatidyl inositol phosphate to inositol triphosphate and diacylglycerol. The former causes release of calcium from the endoplasmic reticulum, while the latter is an activator of Protein Kinase C. Within the PLC family, PLCγ is the only member that contains SH2 and SH3 domains. These domains enable it to interact with receptor tyrosine kinases and become enzymatically activated via phosphorylation. It exists as two isoforms: 1) PLCγ1, which is ubiquitously expressed, and 2) PLCγ2, found primarily in the lymphoid system. PLCγ is essential for growth factor-induced cell motility and mitogenesis. PLCγ1-null mice exhibit retarded embryonic growth and lethality in midgestation. In addition, PDGF stimulation leads to phosphorylation of PLCγ at Tyr 783, and activation of hydrolyzing activity. Overexpression of PLCγ is evident in several forms of cancer and it has been identified as a key mediator of PDGF-dependent cellular transformation. Thus, regulation of PLCγ activity by growth factors is involved in cell growth and transformation.
Western Blotting
Spezifikation
Phospholipase Cg (pY783) | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
PLCγ (pY783) |
27 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Human Phosphorylated PLCγ1 Peptide | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.