Learn More
Phosphotyrosine Monoclonal Antibody (IG2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA4003
Weitere Details : Netto-Gewicht : 0.01000kg
Beinhaltet: 100μg
Beschreibung
MA4-003 detects phosphotyrosine from most species and tissues. MA4-003 has been successfully used in ELISA, Western blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and FACS procedures. This antibody detects proteins which contain phosphorylated tyrosine residues. The MA4-003 antigen is O-phospho-L-tyrosine and O-phospho-DL-tyramine.
Phosphotyrosine Monoclonal antibody specifically detects Phosphotyrosine in Chemical samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
Phosphotyrosine | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide | |
Mouse | |
Affinity chromatography | |
RUO | |
4° C | |
Liquid | |
Chemical |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
IG2 | |
Unconjugated | |
Phosphotyrosine; pTyr; pY | |
O-phospho-L-tyrosine and O-phospho-DL-tyramine | |
100 μg | |
Primary | |
Antibody | |
IgG1 |