missing translation for 'onlineSavingsMsg'
Learn More

PHR1 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA519042

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11307023

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with bovine, human and mouse based on sequence homology.

PHR1 Polyclonal antibody specifically detects PHR1 in Rat samples. It is validated for Western Blot.
Spezifikation

Spezifikation

PHR1
Polyclonal
Unconjugated
Plekhb1
AI315628; evectin-1; Evt1; evt-1; KPL1; MGC128483 protein; PH domain containing protein in retina 1; PH domain containing, retinal 1; PH domain-containing family B member 1; PH domain-containing protein in retina 1; Phr1; PHRET1; pleckstrin homology domain containing B1; pleckstrin homology domain containing, family B (evectins) member 1; pleckstrin homology domain retinal protein 1; Pleckstrin homology domain-containing family B member 1; Plekhb1; retinal 1
Goat
Antigen affinity chromatography
RUO
64471
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9WU68
Plekhb1
Synthetic peptide sequence (PPDSALESPFEE) corresponding to the N-terminus amino acids of PLEKHB1
100 μg
Primary
Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote