missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phycoerythrin Polyclonal Antibody, FITC, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535007
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-35007 targets Phycoerythrin-R/R-PE in FACS and IHC applications and shows reactivity with Tag samples. The PA5-35007 immunogen is highly purified PE.
Phycoerythrin Polyclonal antibody specifically detects Phycoerythrin in Tag samples. It is validated for Flow Cytometry, Immunohistochemistry.Spezifikation
Phycoerythrin | |
Polyclonal | |
FITC | |
PE; RPE; R-PE; R-phycoerythrin | |
highly purified PE | |
100 μg | |
Primary | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Rabbit | |
Protein G | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid | |
Tag |