Learn More
PIK3CB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522152
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-22152 targets PI3 Kinase p110 beta in ICC, IF, and WB applications and shows reactivity with Human and mouse samples. The PA5-22152 immunogen is a recombit fragment corresponding to a region within amino acids 509 and 820 of PI3 kinase p110 beta (Uniprot IDNo.P42338). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Spezifikation
PIK3CB | |
Polyclonal | |
Unconjugated | |
P42338, Q8BTI9, Q9Z1L0 | |
PIK3CB | |
Recombit fragment corresponding to a region within amino acids 509 and 820 of PI3 kinase p110 beta (Uniprot IDNo.P42338) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
PIK3CB | |
P110BETA, PI3K, PI3KBETA, PIK3C1, PI3-kinase p110 subunit beta, PI3-kinase subunit beta, PI3K-beta, PtdIns-3-kinase p110, phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta isoform, ptdIns-3-kinase subunit beta, ptdIns-3-kinase subunit p110-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5291, 74769, 85243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |