missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA515250
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PIK3CD Polyclonal antibody specifically detects PIK3CD in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PIK3CD | |
Polyclonal | |
Unconjugated | |
PIK3CD | |
2410099E07Rik; 2610208K16Rik; APDS; AW545373; catalytic phosphatidylinositol 3-kinase delta; IMD14; p100 delta; p110D; p110delta; phosphatidylinositol 3-kinase catalytic delta polypeptide; phosphatidylinositol 3-kinase catalytic subunit delta; phosphatidylinositol 3-kinase p110 delta; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta; phosphoinositide 3 kinase P110delta; phosphoinositide-3-kinase C; phosphoinositide-3-kinase, catalytic, delta polypeptide; phosphoinositide-3-kinase, catalytic, delta polypeptide variant p37delta; PI3K; PI3K delta; pi3kcd; PI3Kdelta; PI3K-delta; PI3 | |
Rabbit | |
Size-exclusion - Dialysis | |
RUO | |
5293 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O00329 | |
PIK3CD | |
KLH conjugated synthetic peptide between 687-717 amino acids from the C-terminal region of human PI3KCD | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |