missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA515265
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Spezifikation
PIK3R4 | |
Polyclonal | |
Unconjugated | |
Q99570 | |
PIK3R4 | |
KLH conjugated synthetic peptide between 53-83 amino acids from the N-terminal region of human PI3KR4 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PIK3R4 | |
phosphoinositide-3-kinase, regulatory subunit 4, p150 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
30849 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |