Learn More
PI4-Kinase β Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
321.00 € - 650.00 €
Spezifikation
Antigen | PI4-Kinase β |
---|---|
Klon | 7 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15886029
|
BD Biosciences
611816 |
50 μg |
321.00 €
1 Set |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15896029
|
BD Biosciences
611817 |
150 μg |
650.00 €
1 Set |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Phosphoinositide turnover is a well established mechanism of intracellular signal transduction. Sequential phosphorylation of phosphatidylinositol (PtdIns) results inPtdIns-4-phosphate (PIP) and PtdIns-4,5-bisphosphate (PIP2). Phospholipase C (PLC)hydrolyzes PIP2 to inositol-1,4,5-trisphosphate (IP3) which stimulates release of intracellular Ca2+. PIP is generated by phosphorylation of PtdIns at the D4 position of the inositol ring. This event is mediated by the PtdIns 4-kinases (PI4-K). These enzymes are divided into two types (II and III) based on their size and sensitivity to certain compounds. Although the PI4-Ks are abundantly distributed throughout the cell, activity is found primarily in association with membranous structures. Members of this family contain a lipid kinase unique domain and a C-terminal catalytic domain. Two mammalian PI4-Ks, PI4-Kα and PI4-Kβ, have been identified. PI4-Kβ is homologous to the yeast PI4-K, PIK1. Based on its size and sensitivity to wortmanin (a PI3-K inhibitor), PI4-Kβ is classified as a type III enzyme. Although it is found in the cytosol and in association with the Golgi, the specific function of PI4-Kβ is yet to be determined.
Host Species: Mouse
Clone: 7
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human PI4Kβ aa. 411-626
Immunofluorescence, Western Blotting
Spezifikation
PI4-Kinase β | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
7 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Human PI4Kβ aa. 411-626 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.