missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIASy Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141047
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
These sequences are 95% and 100% homologous in mouse and rat. Suggested positive control: liver, Jurkat.
PIASy Polyclonal antibody specifically detects PIASy in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
PIASy | |
Polyclonal | |
Unconjugated | |
PIAS4 | |
e3 sumo- ligase pias4; E3 SUMO-protein ligase PIAS4; FLJ12419; MGC35296; Pias4; Piasg; Pias-gamma; PIASy; protein inhibitor of activated STAT 4; protein inhibitor of activated STAT PIASy; protein inhibitor of activated STAT protein 4; Protein inhibitor of activated STAT protein gamma; protein inhibitor of activated STAT protein PIASy; protein inhibitor of activated STAT, 4; RING-type E3 ubiquitin transferase PIAS4; zinc finger, MIZ-type containing 6; ZMIZ6 | |
Rabbit | |
Protein G | |
RUO | |
362827, 51588, 59004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q8N2W9, Q9JM05 | |
PIAS4 | |
Synthetic peptides corresponding to residues 44-60 and 187-198 of human PIaSg. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |