Learn More
PIG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522197
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (96%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PIG3 Polyclonal antibody specifically detects PIG3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PIG3 | |
Polyclonal | |
Unconjugated | |
TP53I3 | |
p53-induced gene 3 protein; PIG3; quinone oxidoreductase homolog; quinone oxidoreductase PIG3; Quinone oxidoreductase-like protein 2; RGD1304982; TP53I3; tumor protein p53 inducible protein 3; Tumor protein p53-inducible protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q53FA7 | |
TP53I3 | |
Full length human PIG3 Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |