missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGH Monoclonal Antibody (2F8), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA519601
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: TLLIIDSLGI QMTSSYASGK ESTTFIEMGK VKDIVINEAI YMQKVIYYLC ILLKDPVEPH GISQVVPVFQ SAKPRLDCLI EVYRSCQEIL AHQKATSTSP
PIGH Monoclonal antibody specifically detects PIGH in Human samples. It is validated for ELISA, Western Blot.Spezifikation
PIGH | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q14442 | |
PIGH | |
PIGH (NP_004560, 89 a.a. ∽ 188 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2F8 | |
Unconjugated | |
PIGH | |
2210416H01Rik; A930028P05Rik; GPI-H; Mrp64; phosphatidylinositol glycan anchor biosynthesis class H; phosphatidylinositol glycan anchor biosynthesis, class H; phosphatidylinositol glycan, class H; phosphatidylinositol N-acetylglucosaminyltransferase subunit H; Phosphatidylinositol-glycan biosynthesis class H protein; phosphatidylinositol-glycan biosynthesis, class H protein; Pigh; PIG-H | |
Mouse | |
Affinity chromatography | |
RUO | |
5283 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |