missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILRB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542955
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: KLTITQAVTT TTTWRPSSTT TIAGLRVTES KGHSESWHLS LDTAIRVALA Sequence homology: Human: 100%
PILRB Polyclonal antibody specifically detects PILRB in Human samples. It is validated for Western Blot.Spezifikation
PILRB | |
Polyclonal | |
Unconjugated | |
PILRB | |
activating receptor PILRbeta; Activating receptor PILR-beta; cell surface receptor FDFACT; cell surface receptor FDFACT1; cell surface receptor FDFACT2; FDFACT; FDFACT1; FDFACT2; LOC681069; paired immunoglobin-like receptor beta; paired immunoglobin-like type 2 receptor beta; paired immunoglobulin-like receptor beta; paired immunoglobulin-like type 2 receptor beta; PILRB; PP1551; similar to paired immunoglobin-like type 2 receptor beta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UKJ0 | |
PILRB | |
Synthetic peptide directed towards the middle region of human PILRB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |