missing translation for 'onlineSavingsMsg'
Learn More

LIMS1 Monoclonal Antibody (5G7), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA515876

Weitere Details : Netto-Gewicht : 0.10000kg

Artikelnummer. 12897892

  • 461.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-15876 targets PINCH in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human samples. The MA5-15876 immunogen is purified recombinant fragment of human PINCH expressed in E. Coli. . MA5-15876 detects PINCH which has a predicted molecular weight of approximately 37kDa.

LIMS1 Monoclonal antibody specifically detects LIMS1 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

LIMS1
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
P48059
LIMS1
Purified recombinant fragment of human PINCH expressed in E. Coli.
RUO
3987
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
5G7
Unconjugated
LIMS1
2310016J22Rik; 4921524A02Rik; AI507642; AU021743; AW551584; C430041B13Rik; LIM and senescent cell antigen-like domains 1; LIM and senescent cell antigen-like-containing domain protein 1; LIM zinc finger domain containing 1; LIMS1; Lims1l; LIM-type zinc finger domains 1; Particularly interesting new Cys-His protein 1; PINCH; PINCH1; PINCH-1; renal carcinoma antigen NY-REN-48; RGD1560732; senescent cell antigen
Mouse
100 μL
Primary
Human
Antibody
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote