missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINX1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518009
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
PINX1 Polyclonal antibody specifically detects PINX1 in Human samples. It is validated for Western Blot.Spezifikation
PINX1 | |
Polyclonal | |
Unconjugated | |
PINX1 | |
2210403I16Rik; 2610028A01Rik; 67-11-3; 67-11-3 protein; AU024023; EMBL:ABO28828.1}; FLJ20565; hepatocellular carcinoma-related putative tumor suppressor; liver-related putative tumor suppressor; LPTL; Lpts; LPTS1; MGC8850; PIN2 interacting protein 1; PIN2/TERF1 interacting, telomerase inhibitor 1; PIN2/TERF1-interacting telomerase inhibitor 1; PIN2/TRF1-interacting protein; pin2-interact; PIN2-interacting protein 1; PIN2-interacting protein X1; Pinx1; pinx1 {ECO:0000312; Protein 67-11-3; RGD1566025; TRF1-interacting protein 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
54984 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96BK5 | |
PINX1 | |
Synthetic peptide sequence (SMLAERRRKQKWAV) corresponding to the N-terminus amino acids of PINX1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |