missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITPNC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530508
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2. Predicted reactivity: Mouse (97%), Pig (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PITPNC1 Polyclonal antibody specifically detects PITPNC1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PITPNC1 | |
Polyclonal | |
Unconjugated | |
PITPNC1 | |
1110020B03Rik; 5830436L09Rik; AI662802; AI851387; C330017I21Rik; cytoplasmic phosphatidylinositol transfer protein 1; diabetic nephropathy-related protein; Dnr411; Mammalian rdgB homolog beta; mM-rdgBbeta; M-rdgB beta; mrdgBbeta; M-RDGB-beta; phosphatidylinositol transfer protein; phosphatidylinositol transfer protein cytoplasmic 1; phosphatidylinositol transfer protein cytoplasmic 1a; phosphatidylinositol transfer protein, cytoplasmic 1; pitpnc1; pitpnc1a; RDGBB; RDGBB1; RdgBbeta; RDGB-BETA; retinal degeneration B beta; retinal degeneration B beta 1; retinal degeneration B homolog beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26207 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UKF7 | |
PITPNC1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 219 of Human PITPNC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |