missing translation for 'onlineSavingsMsg'
Learn More

PITPNM1 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA518266

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11386233

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with bovine, canine and human based on sequence homology.

PITPNM1 Polyclonal antibody specifically detects PITPNM1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Spezifikation

Spezifikation

PITPNM1
Polyclonal
Unconjugated
PITPNM1
Dres9; drosophila retinal degeneration B homolog; Drosophila retinal degeneration B homolog 1; FLJ44997; membrane-associated phosphatidylinositol transfer protein 1; Mpt1; Mpt-1; NIR2; Nir-2; phosphatidylinositol membrane-associated; phosphatidylinositol transfer protein membrane associated 1; phosphatidylinositol transfer protein, membrane associated 1; phosphatidylinositol transfer protein, membrane-associated 1; Pitpnm; PITPnm 1; PITPNM1; Pyk2 N-terminal domain-interacting receptor 2; R75447; Rd9; RdgB; RDGB1; RDGBA; RDGBA1; retinal degeneration 9; retinal degeneration B alpha 1
Goat
Antigen affinity chromatography
RUO
9600
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O00562
PITPNM1
Synthetic peptide sequence (KARSISLKLDSEE) corresponding to the C-terminus amino acids of PITPNM1
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote