missing translation for 'onlineSavingsMsg'
Learn More

PIWIL1 Recombinant Rabbit Monoclonal Antibody (10H15L2), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marque:  Invitrogen 701177

Informations supplémentaires : Netto-Gewicht : 0.01000kg

Code produit. 15317074

  • 461.00 € / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Description

Description

This antibody is predicted to react with mouse based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

PIWIL1 Recombinant Monoclonal antibody specifically detects PIWIL1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Spécification

Spécification

PIWIL1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1
Recombinant protein corresponding to amino acids 90–213 of human Hiwi
100 μg
Primary
Human, Mouse, Rat
Liquid
Immunocytochemistry, Western Blot
10H15L2
Unconjugated
Q96J94
Rabbit
Protein A
RUO
9271
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions