missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL2 Mouse anti-Mouse, Clone: 11C11, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517208
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control for this product is mouse testis.
The piwi gene of Drosophila belongs to a novel class of evolutionarily conserved genes (the piwi or Argonaute family). The Piwi family‘ is required for germ-and stem-cell development in invertebrates, and two Piwi members, MIWI and MILI (Miwi like; Piwil2; Piwi-like protein 2), are essential for spermatogenesis in mouse. MIWI and MILI partner with Piwi-interacting RNAs (piRNAs) which are 26-31 nucleotides (nt) in length—clearly distinct from the 21-23 nt of microRNAs (miRNAs) or short interfering RNAs (siRNAs). A search for murine small RNAs that might program Piwi proteins for transposon suppression revealed developmentally regulated piRNA loci, some of which resemble transposon master control loci of Drosophila. MILI might be programmed by distinct piRNA populations at different stages of germ cell development, suggesting MILI plays an important role in meiotic prophase progression.Spezifikation
PIWIL2 | |
Monoclonal | |
1 mg/mL | |
HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide | |
Q8CDG1 | |
PIWIL2 | |
Recombit human protein purified from E.coli (GST-mili-1) | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Western Blot | |
11C11 | |
Unconjugated | |
Piwil2 | |
PIWI-L2; PIWI-like 2; PIWIL1L; HILI; MILI | |
Mouse | |
Protein G | |
RUO | |
57746 | |
-20°C | |
Liquid |