Learn More
PKA alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528160
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, Jurkat, NT2D1, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (95%), Dog (100%), Rabbit (97%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKA alpha Polyclonal antibody specifically detects PKA alpha in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PKA alpha | |
Polyclonal | |
Unconjugated | |
PRKACA | |
alternatively spliced; C(s); Calpha; C-alpha-2; C-alpha-S; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit C alpha; cAMP-dependent protein kinase catalytic subunit Calpha1; Cs-PKA; MGC102831; MGC48865; PKA; PKA C-alpha; PKA catalytic subunit alpha; pkac; Pkaca; PKCA1; PKCD; PPNAD4; PRKACA; protein kinase; protein kinase A; protein kinase A alpha; protein kinase A catalytic subunit; protein kinase cAMP-activated catalytic subunit alpha; protein kinase, cAMP dependent, catalytic, alpha; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; sperm cAMP-dependent protein kinase catalytic subunit; sperm cAMP-dependent protein kinase catalytic subunit Cs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18747, 25636, 5566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P05132, P17612, P27791 | |
PRKACA | |
Recombinant fragment corresponding to a region within amino acids 111 and 351 of Human PKA alpha | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |