Learn More
PKA beta Monoclonal Antibody (2G8-1D12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA519698
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: MGNAATAKKG SEVESVKEFL AKAKEDFLKK WENPTQNNAG LEDFERKKTL GTGSFGRVML VKHKATEQYY AMKILDKQKV VKLKQIEHTL NEKRILQAVN FPFLVRLEYA FKDNSNLYMV MEYVPGGEMF SHLRRIGRFS EPHARFYAAQ IVLTFEYLHS LDLIYRDLKP ENLLIDHQGY IQVTDFGFAK RVKGRTWTLC GTPEYLAPEI ILSKGYNKAV DWWALGVLIY EMAAGYPPFF ADQPIQIYEK IVSGKNF
PKA beta Monoclonal antibody specifically detects PKA beta in Human samples. It is validated for ELISA.Spezifikation
PKA beta | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P22694 | |
PRKACB | |
PRKACB (AAH16285, 1 a.a. ∽ 257 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
2G8-1D12 | |
Unconjugated | |
PRKACB | |
cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit beta; PKA C-beta; Pkacb; PRKACB; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit beta; protein kinase, cAMP dependent, catalytic, beta; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, beta | |
Mouse | |
Affinity chromatography | |
RUO | |
5567 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |