missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKAR1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA515368
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PRKAR1A Polyclonal antibody specifically detects PRKAR1A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PRKAR1A | |
Polyclonal | |
Unconjugated | |
PRKAR1A | |
1300018C22Rik; ACRDYS1; ADOHR; cAMP-dependent protein kinase regulatory subunit RIalpha; cAMP-dependent protein kinase type I-alpha regulatory chain; cAMP-dependent protein kinase type I-alpha regulatory subunit; cAMP-dependent protein kinase type I-alpha regulatory subunit, N-terminally processed; CAR; Carney complex type 1; CNC; CNC1; PKR1; PPNAD1; PRKAR1; Prkar1a; protein kinase A type 1a regulatory subunit; protein kinase cAMP-dependent type I regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type 1, alpha; protein kinase, cAMP dependent regulatory, type I, alpha; protein kinase, cAMP-dependent, regulatory subunit type I alpha; protein kinase, cAMP-dependent, regulatory, type I, alpha; RIalpha; RIIA; Tissue-specific extinguisher 1; Tse1; Tse-1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5573 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P10644 | |
PRKAR1A | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human Protein Kinase A regulatory subunit I alpha | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |