Learn More
PKC gamma Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527732
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Molt-4, mouse brain, rat brain. Predicted reactivity: Mouse (98%), Rat (98%), Rabbit (98%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKC gamma Polyclonal antibody specifically detects PKC gamma in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Spezifikation
PKC gamma | |
Polyclonal | |
Unconjugated | |
PRKCG | |
PKC; Pkcc; PKCG; PKCgamma; PKC-gamma; PKCI; Prkc; Prkcc; PRKCG; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, gamma; RATPKCI; SCA14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18752, 24681, 5582 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
0.55 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P05129, P63318, P63319 | |
PRKCG | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 166 of Human PKC gamma | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |