missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA130288
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HL60 cell lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PRKD3 Polyclonal antibody specifically detects PRKD3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PRKD3 | |
Polyclonal | |
Unconjugated | |
Prkd3 | |
4930557O20Rik; 5730497N19Rik; EPK2; nPKC-D1; nPKC-mu; nPKC-nu; PKCM; PKC-MU; Pkcnu; PKC-NU; PKD; PKD1; PKD3; PRKCM; Prkcn; PRKD1; Prkd3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D3; protein kinase EPK2; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D3 | |
Rabbit | |
Protein G | |
RUO | |
23683, 75292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
O94806, Q8K1Y2 | |
Prkd3 | |
KLH conjugated synthetic peptide between 352-384 amino acids from human PKC-nu | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |