Learn More
PKC theta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529089
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (80%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKC theta Polyclonal antibody specifically detects PKC theta in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PKC theta | |
Polyclonal | |
Unconjugated | |
PRKCQ | |
A130035A12Rik; AW494342; EC 2.7.11.13; kinase nPKC-theta; KPCT; MGC126514; MGC141919; nPKC-theta; PKC theta; PKC-0; Pkcq; PKCtheta; PKC-theta; PRKCQ; PRKCT; protein kinase C theta; protein kinase C theta type; protein kinase C, theta; RP11-563J2.1; si:ch211-239o15.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18761, 5588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q02111, Q04759 | |
PRKCQ | |
Recombinant fragment corresponding to a region within amino acids 15 and 294 of Human PKC theta | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |