missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKD1L3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSP00014W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water.
Spezifikation
PKD1L3 | |
Polyclonal | |
PKD1L3 | |
PKD1L3, PK1L3, Polycystin-1L3, PC1-like 3 protein | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q7Z443 | |
PKD1L3 | |
A synthetic peptide from the 1t extracellular loop of human Polycystic kidney disease protein 1-like 3 | |
RUO | |
342372 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |