Learn More
PRKD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA123446
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
According to customer data, this antibody can detect pure human PKD2, endogenous PKD2 in resting neonatal rat cardiac fibroblasts, and it reacts weakly in resting neonatal rat cadiomyocytes. However, it does not detect PKD2 in PMA stimulated rat cells or pure phosphorylated PKD2.
PRKD2 Polyclonal antibody specifically detects PRKD2 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
PRKD2 | |
Polyclonal | |
Unconjugated | |
PRKD2 | |
AI325941; HSPC187; nPKC-D2; PKD2; Prkd2; protein kinase D2; RGD1308054; serine/threonine-protein kinase D2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25865 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9BZL6 | |
PRKD2 | |
Purified, full-length native human protein kinase D2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |