missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA130299
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa cell lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKN3 Polyclonal antibody specifically detects PKN3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PKN3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6P5Z2 | |
PKN3 | |
KLH conjugated synthetic peptide between 463-492 amino acids from the C-terminal region of human PKN beta. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PKN3 | |
AW209115; BC034126; Pkn3; PKNBETA; protein kinase N3; protein kinase PKNbeta; protein kinase PKN-beta; protein-kinase C-related kinase 3; Serine/threonine-protein kinase N3; UTDP4-1 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
29941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |