missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518800
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine based on sequence homology.
PLA2 Polyclonal antibody specifically detects PLA2 in Human samples. It is validated for Western Blot.Spezifikation
PLA2 | |
Polyclonal | |
Unconjugated | |
PLA2G1B | |
Group IB phospholipase A2; group IB secretory phospholipase A(2); MGC119834; MGC119835; phosphatidylcholine 2-acylhydrolase 1B; phospholipase; phospholipase A2; phospholipase A2 group IB; phospholipase A2, group 1B; phospholipase A2, group IB (pancreas); phospholipase A2, group IB, pancreas; phospholipase A2, major isoenzyme; PLA2; PLA2A; Pla2g1b; PLA2-Ib; PPLA2; sPLA(2)-IB; sPLA2IB; unnamed protein product | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5319 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P04054 | |
PLA2G1B | |
Synthetic peptide sequence (NKAHKNLDTKKYCQS) corresponding to the C-terminus amino acids of PLA2G1B | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |