missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G2A Monoclonal Antibody (620501), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524278
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, no cross-reactivity with recombinant human PLA2G1B, G2A, or G7 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
PLA2G2A Monoclonal antibody specifically detects PLA2G2A in Human samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
PLA2G2A | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P14555 | |
PLA2G2A | |
Sf 21-derived recombinant human PLA2G2A Asn21-Cys144 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunoprecipitation, Western Blot | |
620501 | |
Unconjugated | |
PLA2G2A | |
EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1; non-pancreatic secreted type II phospholipase A2; non-pancreatic secretory phospholipase A2; NPS-PLA2; phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2 group IIA; phospholipase A2, group IIA (platelets, synovial fluid); phospholipase A2, membrane associated; Pla2; PLA2B; PLA2BPLA2; PLA2G2A; PLA2L; PLA2LPLA2S; PLA2S; PLAS1; platelet phospholipase A2; RASF-A; secretory group II phospholipase A2; sPLA2; sPla2-IIA | |
Mouse | |
Protein A/G | |
RUO | |
5320 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |