Learn More
PLA2G4F Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531735
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Human (99%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLA2G4F Polyclonal antibody specifically detects PLA2G4F in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLA2G4F | |
Polyclonal | |
Unconjugated | |
PLA2G4F | |
3.1.1.4; 4732472I07Rik; cPLA2-zeta; cytosolic phospholipase A2 zeta; Gm647; Phospholipase A2 group IVF; phospholipase A2, group IVF; Pla2g4f; PLA2G4FZ; Pla2zeta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255189 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q68DD2 | |
PLA2G4F | |
Recombinant protein fragment corresponding to a region within amino acids 379 and 602 of Human PLA2G4F | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |