missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA526936
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PLA2G6 Polyclonal antibody specifically detects PLA2G6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLA2G6 | |
Polyclonal | |
Unconjugated | |
PLA2G6 | |
85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; BB112799; CaI-PLA2; Group VI phospholipase A2; GVI; GVI PLA2; INAD1; intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA(2)beta; iPLA2; iPLA2beta; iPLA2-beta; IPLA2-VIA; NBIA2; NBIA2A; NBIA2B; neurodegeneration with brain iron accumulation 2; PARK14; patatin-like phospholipase domain-containing protein 9; phospholipase A2 group VI; phospholipase A2, group VI; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2; PLA2G6; PLPLA9; PNPLA9 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
8398 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
O60733 | |
PLA2G6 | |
KLH conjugated synthetic peptide between 551-580 amino acids from the central region of human PLA2G6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |