missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA526819
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PLA2G7 Polyclonal antibody specifically detects PLA2G7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLA2G7 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q13093 | |
PLA2G7 | |
KLH conjugated synthetic peptide between 200-228 amino acids from the central region of human PLA2G7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PLA2G7 | |
1-alkyl-2-acetylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase; Group-VIIA phospholipase A2; gVIIA-PLA2; LDL-associated phospholipase A2; LDL-PLA(2); LDL-PLA2; lipoprotein-associated phospholipase A2; LP-PLA2; PAF 2-acylhydrolase; PAF acetylhydrolase; PAFAD; Pafah; phospholipase A2 group VII; phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma); PLA2G7; Platelet-activating factor acetylhydrolase; R75400 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
7941 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |