missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2R1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519141
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine and porcine based on sequence homology.
PLA2R1 Polyclonal antibody specifically detects PLA2R1 in Human samples. It is validated for Flow Cytometry.Spezifikation
PLA2R1 | |
Polyclonal | |
Unconjugated | |
Pla2r1 | |
180 kDa secretory phospholipase A2 receptor; CLEC13C; C-type lectin domain family 13 member C; mannose receptor, C type 2; Mrc2; M-type receptor; phospholipase A2 receptor 1; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; phospholipase A2, group IB, pancreas, receptor; Pla2g1br; PLA2G1R; PLA2-I receptor; PLA2IR; Pla2r; Pla2-r; PLA2R1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22925 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q13018 | |
Pla2r1 | |
Synthetic peptide sequence (EKKKDTPKQHGT) corresponding to the internal amino acids of PLA2R1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |