Learn More
PKP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529098
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431. Predicted reactivity: Mouse (96%), Rat (96%), Dog (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKP1 Polyclonal antibody specifically detects PKP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PKP1 | |
Polyclonal | |
Unconjugated | |
PKP1 | |
AI528551; armadillo; B6P; band 6; band 6 polypeptide B6P; band 6 protein; Band-6 protein; MGC138829; Pkp1; plakophilin; plakophilin 1; plakophilin 1 (ectodermal dysplasia/skin fragility syndrome); plakophilin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13835 | |
PKP1 | |
Recombinant protein fragment corresponding to a region within amino acids 184 and 483 of Human Plakophilin 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |