missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKP3 Monoclonal Antibody (7F6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA518080
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This target has a predicted molecular weight of 87 kDa.
PKP3 Monoclonal antibody specifically detects PKP3 in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
PKP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9QY23, Q9Y446 | |
PKP3 | |
Recombinant fragment corresponding to amino acids 1-308 of Human Plakophilin 3 fused to maltose binding protein | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
7F6 | |
Unconjugated | |
PKP3 | |
2310056L12Rik; Pkp3; plakophilin 3; plakophilin 3b; plakophilin-3 | |
Mouse | |
Protein A | |
RUO | |
11187, 56460 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |